Chloroacetic acid 99%, flake

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Monochloroacetic acid

108510010 108510025 108510040 108512500
TS108510010EA 68 USD
TS108510010 TS108510025 TS108510040 TS108512500
Chloroacetic acid 99%, flake
Chloroacetic acid
Formula: ClCH₂COOH
MW: 94.5 g/mol
Boiling Pt: 188
Melting Pt: 61…63 °C
Density: 1.58 g/cm³
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002683
CAS Number: 79-11-8
EINECS: 201-178-4
UN: 1751
ADR: 6.1 II
Merck Index: 13,02129

Order Now

Specification Test Results

Appearance White Crystalline flakes
Infrared spectrum Conforms
Melting point 61 to 63 °C
Water ≤0.3 %
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR