Tetrachloroethylene 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene

138010010 138010025 138010100 138015000
TS138010010EA 101 USD
TS138010010 TS138010025 TS138010100 TS138015000
Tetrachloroethylene 99%, Extra Pure
Tetrachloroethylene
Formula: C₂Cl₄
MW: 165.83 g/mol
Boiling Pt: 120…122 °C (760 mmHg)
Melting Pt: –22 °C
Density: 1.625 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000834
CAS Number: 127-18-4
EINECS: 204-825-9
UN: 1897
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.01 %
GC ≥98.5 %
Color scale ≤15 APHA
Residue after evaporation ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR