Amberlite® FPC3500 ion-exchange resin

Supplier: Thermo Scientific Chemicals

389990010 389992500
TS389990010EA 157 USD
TS389990010 TS389992500
Amberlite® FPC3500 ion-exchange resin
Amberlite® FPC3500
Storage Temperature: Ambient
MDL Number: MFCD08460528
CAS Number: 950182-46-4

Order Now

Specification Test Results

Appearance White to light yellow Beads
Particle size ≤5.0 % (> 1.180 mm)
Harmonic mean size 0.450 to 0.650 mm
Uniformity coefficient ≤2.00
Moisture holding capacity 60 to 70 % (H)
Total volume capacity ≥2.60 eq/l (H)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR