Amberlite® IRA-410 (Cl) resin_ion-exchange

Supplier: Thermo Scientific Chemicals

Synonyms: Amberlite® IRA-410 Cl type

202291000 202295000
TS202291000EA 21 USD
TS202291000 TS202295000
Amberlite® IRA-410 (Cl) resin_ion-exchange
Amberlite® IRA-410 (Cl)
Storage Temperature: Ambient
MDL Number: MFCD00132710
CAS Number: 9002-26-0

Order Now

Specification Test Results

Appearance Transparent light yellow to light orange Beads
Particle size ≤5.0 % (>1.180 mm)
Harmonic mean size 0.600 to 0.750 mm
Uniformity coefficient ≤1.60
Moisture holding capacity 45.0 to 51.0 % (Cl-)
Total volume capacity ≥1.25 eq/l (Cl-)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR