Stoddard solvent

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mineral spirits

419730050 419730010
TS419730050EA 498 USD
TS419730050 TS419730010
Stoddard solvent
Stoddard solvent
Boiling Pt: 149…213 °C (760 mmHg)
Melting Pt: –40 °C
Density: 0.787
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00132767
CAS Number: 8052-41-3
UN: 1300
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear liquid
Boiling point 149 to 213°C (ASTM D 86)
Density (15.6°C) 754 to 820 kg/m³ (ASTM D 4052)
Additional info Kauri butanol value: 29 to 45 (ASTM D 1133)
Color scale ≤25 APHA (ASTM D 1209)
Bromine number ≤5 g Br/100 g (ASTM D 1159)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR