Hydrochloric acid 25% in water, pure

Supplier: Thermo Scientific Chemicals
Danger

387790010 387790025
TS387790010EA 37 USD
TS387790010 TS387790025
Hydrochloric acid 25% in water, pure
Hydrochloric acid
Formula: HCl
Boiling Pt: 107 °C (760 mmHg)
Melting Pt: –70 °C
Density: 1.12 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Residue after ignition ≤0.002 %
Titration with NaOH ≥25.0 %
Sulfate (SO4) ≤0.002 %
Iron (Fe) ≤0.0001 %
Heavy metals (as Pb) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR