2-Mercaptopyridine-N-oxide sodium salt 40% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Mercaptopyridinen-oxide sodiumsalt, 2-Mercaptopyridine N-oxide sodium

257740010 257740025 257740050
TS257740010EA 517 USD
TS257740010 TS257740025 TS257740050
2-Mercaptopyridine-N-oxide sodium salt 40% (w/w) in aqueous solution
2-Mercaptopyridine N-oxide sodium salt
Formula: C₅H₄NNaOS
MW: 149.15 g/mol
Melting Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD01941547
CAS Number: 3811-73-2
EINECS: 223-296-5
UN: 3082
ADR: 9 III

Order Now

Specification Test Results

Appearance Yellow to amber Solution
pH 8.5 to 10.5 (10 % at 20°C)
Titration with HClO4 40 to 42 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR