Molecular sieve A4 (0.4 nm, 4 Å), powder <50 µm
Supplier: Thermo Scientific Chemicals
214800010
214800025
214800100
214800250
214805000
TS214800010EA
153
USD
TS214800010
TS214800025
TS214800100
TS214800250
TS214805000
Molecular sieve A4 (0.4 nm, 4 Å), powder <50 µm
Storage Temperature:
Ambient |
MDL Number:
MFCD03457537 CAS Number: 70955-01-0 |
Specification Test Results
Appearance | White to beige Powder |
pH | 9 to 12 (10 % suspension in water) |
Residual water | ≤3 % |
Water adsorption | ≥18 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...