tert-Butanol ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol

390690010 390690025
TS390690010EA 110 USD
TS390690010 TS390690025
tert-Butanol ACS
tert-Butanol
Formula: (CH₃)₃COH
MW: 74.12 g/mol
Boiling Pt: 82.2
Melting Pt: 25.3
Density: 0.78 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3 II
Merck Index: 12,01577

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.1 % (K.F.)
GC ≥99.0 %
Color scale ≤20 APHA
Residue after evaporation ≤0.003 %
Carbonyl compounds ≤0.01 % (as formaldehyde)
Titrable acid ≤0.001 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR