Cyclopentane, extra dry 95+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pentamethylene

397680010 397681000
TS397680010EA 197 USD
TS397680010 TS397681000
Cyclopentane, extra dry 95+%, AcroSeal™
Cyclopentane
Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 49
Melting Pt: -93.3
Density: 0.75 g/cm³
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00001356
CAS Number: 287-92-3
UN: 1146
ADR: 3 II
Merck Index: 13,02769

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥95.0 %
Residue after evaporation ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR