Nitrobenzene 99.5% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
447650010
447652500
TS447650010EA
155
USD
TS447650010
TS447652500
Nitrobenzene 99.5% for analysis
Nitrobenzene
Formula:
C₆H₅NO₂ MW: 123.11 g/mol Boiling Pt: 210.8 Melting Pt: 5.7 Density: 1.205 g/cm³ Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007043 CAS Number: 98-95-3 UN: 1662 ADR: 6.1 II Merck Index: 13,06621 |
Specification Test Results
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5520 to 1.5540 (20°C, 589 nm) |
Water | ≤0.15 % |
GC | ≥99.4 % |
Residue after evaporation | ≤0.005 % |
Chloride (Cl) | ≤5 ppm |
Cadmium (Cd) | ≤0.05 ppm |
Calcium (Ca) | ≤0.5 ppm |
Chromium (Cr) | ≤0.02 ppm |
Cobalt (Co) | ≤0.02 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.1 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Manganese (Mn) | ≤0.02 ppm |
Nickel (Ni) | ≤0.02 ppm |
Zinc (Zn) | ≤0.1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...