qPCR Master Mix Kit
Supplier: STEMCELL Technologies
07516 07517
MSPP-07516EA 204.62 USD
MSPP-07516 MSPP-07517
qPCR Master Mix Kit
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
For probe-based assays and arrays.
  • Efficient, sensitive, and reproducible
  • Optimal performance when using standard or fast cycling conditions
  • Ideal for high-throughput applications and overnight experiments
  • Compatible with various real-time qPCR instruments
qPCR Master Mix is a 2X concentrated solution optimized for TaqMan® probe-based, real-time quantitative polymerase chain reaction (qPCR). It is intended for use in combination with gene-specific primers (to amplify target cDNA) and fluorogenic-labeled probes that use the 5' nuclease activity of Taq DNA polymerase to produce a fluorescent signal. The rate of accumulation of the fluorescent signal is used to quantify the cDNA, and thereby to determine the amount of mRNA present in the original sample. qPCR Master Mix includes a Hot Start DNA polymerase, dNTPs, MgCl₂, enhancers, and stabilizers. The qPCR Master Mix Kit also includes ROX Reference Dye as a separate component, making it compatible with both reference dye-dependent and -independent qPCR systems.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').