Tolcapone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 3,4-Dihydroxy-4'-methyl-5-nitrobenzophenone

T3856-200MG T3856-50MG
TCT3856-200MGEA 356.38 USD
TCT3856-200MG TCT3856-50MG
Tolcapone ≥98.0% (by HPLC)
Tolcapone
Formula: C₁₄H₁₁NO₅
MW: 273.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00866569
CAS Number: 134308-13-7
Merck Index: 14,09508

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 142.0 to 146.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR