Klinger BlackBoard Optics™ Basic Set (Magnetic)
Supplier: KLINGER EDUCATION PROD CORP SE
Klinger BlackBoard Optics™
KO4100M
470006-930EA 1395 USD
470006-930
Klinger BlackBoard Optics™ Basic Set (Magnetic)
Educational Materials Physics Educational Materials Optics Learning Activities
Ray-projectors, lenses, and mirrors that mount directly to the chalkboard, via magnet clamps.
  • Perform experiments directly on the board, chalking in lines, measuring angles, and calculating formulas
  • Packaged in a durable cardboard box with custom cut foam for easy and safe storage
Klinger Blackboard Optics™ is a unique approach to teaching principles of optics and properties of light. Experiments in refraction and reflection are performed directly on the blackboard without darkening the room. Visibility is assured because of the large acrylic lenses. The special ray projectors may be arranged to form a pencil beam of light or a divergent cone by means of a fingertip adjustment. With the components in place, the instructor can chalk in axes, normals, tangents, and equations directly on the chalkboard.
Delivery information: Kit Includes: (2) single ray projectors, (2) projector magnetic holders, (1) planoconvex lens, (1) planoconcave lens, (1) semicircular lens, (1) parallel sided block, (1) right angle prism, and (2) plane mirrors.