Anti-IgG Goat Polyclonal Antibody

Supplier: AAT BIOQUEST INC
50301 50302
77462-848EA 110.32 USD
77462-848 77462-850
Anti-IgG Goat Polyclonal Antibody
Antibodies
Anti-Human IgG (Fc specific) antiserum is produced in goat using purified human IgG, Fc fragment, as the immunogen.

  • No reactivity is observed with the Fab fragment of human IgG, light chains, IgA, or IgM
  • No interspecies cross reactivity with mouse or rat serum proteins
  • Purified through affinity chromatography

The antibody preparation is specific for human IgG, Fc fragment when tested against purified human IgA, IgG (Fc and Fab fragments).

Caution: Research use only.

Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Fc)
Reactivity: (Fc)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR