Chloroform-[D₁] 100.0 atom % D for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chloroform-D
, Trichloromethane-D1
, Chloroform-[²H₁]
320680075
TS320680075EA
77.3
USD
TS320680075
Chloroform-[D₁] 100.0 atom % D for NMR spectroscopy
Chloroform-[D1]
Packaged in 0.75 ml ampoules
Formula:
CDCl₃ MW: 120.38 g/mol Boiling Pt: 60.8 °C (1013 hPa) Melting Pt: –64 °C Density: 1.5 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000827 CAS Number: 865-49-6 EINECS: 212-742-4 UN: 1888 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.01 % (Coulometric) |
D-Enrichment | ≥99.95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...