EasySep™ EasyStand™ Linkable Magnet Stands with Magnets
Supplier: STEMCELL Technologies
18131 18135 18137
MSPP-18131EA 2031.54 USD
MSPP-18131 MSPP-18135 MSPP-18137
EasySep™ EasyStand™ Linkable Magnet Stands with Magnets
Magnets
Linkable stands are available in one, four, or six piece configurations. Each kit is supplied with 1, 4, or 6 EasySep™ magnets
  • Simplify the separation of one or more samples at one time
  • Magnets included
The EasySep™ EasyStand™ facilitates the separation of one or more samples at one time. Each EasySep™ EasyStand™ can hold a single EasySep™ magnet, and a simple system links up to 6 individual EasySep™ EasyStands™ together. After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').