Human TrkA Luciferase Reporter Assay Kit
Supplier: INDIGO BIOSCIENCES INC MS
IB27011 IB27012
104087-904KT 1163.08 USD
104087-904 104087-906
Human TrkA Luciferase Reporter Assay Kit
Assays Enzyme Assays
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO's tropomyosin receptor kinase A reporter assay allows you to quickly screen test samples to quantify functional activity, either agonist or antagonist, they may exert against TrkA (NTRK1).
  • Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
  • Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, luciferase detection reagent, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)
This is an all-inclusive cell-based luciferase reporter assay kit targeting the human tropomyosin receptor kinase A (TrkA). INDIGO's TrkA reporter assay utilizes proprietary mammalian cells that have been engineered to provide constitutive expression of the TrkA. In addition to TrkA reporter cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, a reference agonist, luciferase fetection reagent, and a cell culture-ready assay plate. The principal application of this assay is in the screening of test samples to quantify any functional activity, either agonist or antagonist, that they may exert against TrkA. This kit provides researchers with: Clear, reproducible results, exceptional cell viability post-thaw. Consistent results lot to lot.
Ordering information: Kits are all-inclusive assay systems featuring live cells.
Delivery information: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C. Do not store in liquid nitrogen.