Repositionable Vinyl Labels for BradyJet™ J7300 Color Label Printer
Supplier: Brady Worldwide
170455 170456 170457
77581-092RL 176.89 USD
77581-092 77581-024 77580-948
Repositionable Vinyl Labels for BradyJet™ J7300 Color Label Printer
Labels
Specialized aggressive adhesive allows sticking, removing, and repositioning labels with ease, without leaving behind adhesive residue or surface damage.
  • Repositionable acrylic adhesive
  • Matte finish
  • Service temperature range: –40 to +140 °F
Durable, flexible material is purpose-built for areas where identification needs change frequently. Ideal for Lean/5S and work in process labeling as well as warehousing racks, bins, and storage. Works well in low temperatures with a service temperature of –40 to +140 °F (–40 to +60 °C). Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 colour Label Printer. Ink cartridges are required to print on this material (use part numbers P-CMY-1 and P-K-1).