Brady® Chemical and Temperature Resistant Polypropylene Labels
Supplier: Brady Worldwide
170846 170847
76796-008EA 173.3 USD
76796-008 76796-098
Brady® Chemical and Temperature Resistant Polypropylene Labels
Labels
Specialized labels are pre-sized for use on dry, room-temperature specimen containers or vials.
  • Permanent acrylic adhesive
  • Recommended ribbon: R4300 (94000-380)
Durable labels are resistant to extreme temperatures, moderate chemical contact, and everyday use. Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces in extreme conditions.
Certifications: RoHS compliant to 2011/65/EU (Pass 2015/863).