Jumbo Sack Samplers
Supplier: SAMPLING SYSTEMS LTD
1309A-400 1309A-800 1309B-400 1309B-800 1309C-400 1309C-800
77576-220EA 214.21 USD
77576-220 77576-222 77576-224 77576-226 77576-228 77576-230
Jumbo Sack Samplers
Samplers Solid Samplers
The jumbo sack sampler is designed for sampling large granules from sacks.
  • Ideal for sampling granules from bags on pallets
  • Can sample larger granules up to 10 mm in size
  • Simple to use
  • Ideal for most powders and granules
  • Available from stock
  • Fast delivery
The jumbo aack sampler is made from 316L Stainless steel, and is perfect for sampling powders and granules in sacks. There are various models to suit specific applications, including a model with a plain end which the user can attach to their own bag. A model with an end cap allows the user to retain the sample in the tube. Additionally, a model is available with an adaptor to enable the user to collect the sample in a bottle.