SteriWare® Powder Spatulas, Disposable
Supplier: SAMPLING SYSTEMS LTD
8095B-150 8095B-150S 8095B-250 8095B-250S
77577-034EA 141.56 USD
77577-034 77577-036 77577-038 77577-040
SteriWare® Powder Spatulas, Disposable
Spatulas
PP. These disposable spatulas feature a rounded handle and a robust blade.
  • Manufactured and packed in a controlled atmosphere (class 100000 cleanroom)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • Approved for use in ATEX Zones 0, 1 and 2 (contact Sampling Systems for full details)
  • Disposable: No cleaning is required before or after sampling
  • Available as gamma irradiated sterile (SAL 10⁻⁶)
Suitable for sterile sampling and dispensing quantities of powders in all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.).