Buckets with Spout, Stainless Steel
Supplier: SAMPLING SYSTEMS LTD
A686-15 A686-20
77578-106EA 521.14 USD
77578-106 77578-110
Buckets with Spout, Stainless Steel
Buckets
These buckets with spouts make emptying fast and simple.
  • Designed for heavy duty use
  • Perfect for use in pharma, chemical and food
  • Made from chemically resistant 316L stainless steel
  • Optional lid available
The Sampling Systems range of buckets with spouts are designed with a handy pouring spout which makes emptying fast and simple. Made from chemically resistant 316L stainless steel, they can be widely used across many applications in pharma, chemical and food.