Ethyl 2-Oxocycloheptanecarboxylate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 2-Oxocycloheptanecarboxylic acid Ethyl ester

E1056-200MG E1056-1G
TCE1056-200MGEA 84 USD
TCE1056-200MG TCE1056-1G
Ethyl 2-Oxocycloheptanecarboxylate ≥95.0% (by GC)
Ethyl 2-Oxocycloheptanecarboxylate
Formula: C₁₀H₁₆O₃
MW: 184.24 g/mol
Boiling Pt: 126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00604086
CAS Number: 774-05-0

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR