N-[4-(2,4-Dimethylphenyl)-2-thiazolyl]-4-pyridinecarboxamide ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N-[4-(2,4-Dimethylphenyl)thiazol-2-yl]isonicotinamide, Nek2/Hec1 inhibitor

D5563-10MG D5563-50MG
TCD5563-10MGEA 147 USD
TCD5563-10MG TCD5563-50MG
N-[4-(2,4-Dimethylphenyl)-2-thiazolyl]-4-pyridinecarboxamide ≥98.0% (by HPLC)
N-[4-(2,4-Dimethylphenyl)-2-thiazolyl]-4-pyridinecarboxamide
Formula: C₁₇H₁₅N₃OS
MW: 309.39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03384899
CAS Number: 560103-80-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 13.00 to 14.20 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR