Desoximetasone ≥97.0% (by HPLC)

Supplier: TCI America
Warning

D5515-250MG D5515-50MG
TCD5515-250MGEA 407 USD
TCD5515-250MG TCD5515-50MG
Desoximetasone ≥97.0% (by HPLC)
Desoximetasone
Formula: C₂₂H₂₉FO₄
MW: 376.47 g/mol
Melting Pt: 217 °C
MDL Number: MFCD00083301
CAS Number: 382-67-2
EINECS: 206-845-3
Merck Index: 14,02930

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Specific rotation [a]20/D +115 to +119 deg (C=1,CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR