Triisopropylsilyl acrylate 98,0 GC_ASSAY_METHOD Stabilised
Supplier: TCI America
|
Warning
|
Synonyms:
Acrylic acid triisopropylsilyl ester
Stabilization:
BHT
T3789-25G
T3789-5G
TCT3789-25GEA
205
USD
TCT3789-25G
TCT3789-5G
Triisopropylsilyl acrylate 98,0 GC_ASSAY_METHOD Stabilised
Triisopropylsilyl acrylate
Formula:
C₁₂H₂₄O₂Si MW: 228.41 g/mol Boiling Pt: 102 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01861733 CAS Number: 157859-20-6 EINECS: 457-670-6 UN: 3082 ADR: 9 III |
Specification Test Results
Appearance | Colourless to Almost Colourless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...