Zinc naphthenate (∼8% Zn)
Supplier: TCI America
|
Danger
|
Z0039-25ML
Z0039-500ML
TCZ0039-25MLEA
23
USD
TCZ0039-25ML
TCZ0039-500ML
Zinc naphthenate (∼8% Zn)
Zinc naphthenate
Formula:
2(C₁₁H₇O₂)·Zn Flash Pt: 43 °C |
MDL Number:
MFCD00147699 CAS Number: 12001-85-3 EINECS: 234-409-2 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Yellow to Brown clear liquid |
Zinc | 7.5 to 8.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...