1,2-Bis(4-bromophenyl)ethane ≥98.0% (by GC)

Supplier: TCI America

B5853-1G B5853-5G
TCB5853-1GEA 81 USD
TCB5853-1G TCB5853-5G
1,2-Bis(4-bromophenyl)ethane ≥98.0% (by GC)
1,2-Bis(4-bromophenyl)ethane
Formula: C₁₄H₁₂Br₂
MW: 340.06 g/mol
Boiling Pt: 198 °C
MDL Number: MFCD00634290
CAS Number: 19829-56-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR