AGK2 ≥97.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Cyano-3-[5-(2,5-dichlorophenyl)furan-2-yl]-N-(quinolin-5-yl)acrylamide, 2-Cyano-3-(5-(2,5-dichlorophenyl)furan-2-yl)-N-(quinolin-5-yl)acrylamide
A3193-100MG
A3193-25MG
A3193-5MG
TCA3193-100MGEA
443
USD
TCA3193-100MG
TCA3193-25MG
TCA3193-5MG
AGK2 ≥97.0% (by HPLC)
AGK2
Formula:
C₂₃H₁₃Cl₂N₃O₂ MW: 434.28 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01909444 CAS Number: 304896-28-4 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Elemental analysis (Nitrogen) | 9.40 to 10.20 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...