4,6-Dichloronicotinic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 4,6-dichloropyridine-3-carboxylic acid

D5311-1G D5311-5G
TCD5311-1GEA 31 USD
TCD5311-1G TCD5311-5G
4,6-Dichloronicotinic acid ≥98.0% (by titrimetric analysis)
4,6-Dichloronicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192.00 g/mol
MDL Number: MFCD00234146
CAS Number: 73027-79-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR