2-Amino-6-chloro-3-nitropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 6-Chloro-3-nitro-2-pyridylamine, 6-Chloro-3-nitropyridin-2-amine

A1756-25G A1756-5G
TCA1756-25GEA 90 USD
TCA1756-25G TCA1756-5G
2-Amino-6-chloro-3-nitropyridine ≥98.0% (by GC)
2-Amino-6-chloro-3-nitropyridine
Formula: C₅H₄ClN₃O₂
MW: 173.56
MDL Number: MFCD00044350
CAS Number: 27048-04-0
EINECS: 248-188-5

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 195.0 to 199.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR