Saturix Cleanroom Copy Papers
Supplier: FG CLEAN WIPES TE
97177YW8A00 97177WE8A00 97177WEAG00 97177BEAG00 97177PK8A00 97177YW8A00 97177GN8A00
76238-324CS 465.34 USD
76238-324 76238-326 76239-984 76241-202 76244-876 76244-882 76244-888
Saturix Cleanroom Copy Papers
Paper
Specially designed for use in cleanrooms, each page of copy paper is impregnated and coated with a unique polymer formulation that inhibits particulate generation and chemical extractables.
  • Special anti-contaminant polymer formulation
  • Copy paper compatible with wet and dry systems
Designed for use in ISO Class 3 or higher environments, this cleanroom copy paper is suitable for applications in pharmaceutical and compounding pharmaceutical, biotech, medical device manufacturing, and semiconductor manufacturing industries.