Corning® FiltrEX™ 96-Well Filter Plates, Corning
Supplier: Corning
3508 3504 3510 3511 3505
46610-070CS 1385.89 USD
46610-070 77776-306 77776-308 77776-310 77776-456
Corning® FiltrEX™ 96-Well Filter Plates, Corning
Microplates Filter Plates
FiltrEX™ filter plates meet the industry standards for plate dimensions. The rigid side walls make the plate ideal for automation and the wide skirt accepts barcodes. Individual filter discs are encapsulated in the plate by a process that ensures 100% integrity of each well. The design of the nozzle prevents sample wicking and cross-contamination. Glass fibre filter plates can be used for a variety of applications, such as plasmid isolation, DNA purification, PCR clean-up or receptor/ligand binding assays. The low-binding hydrophilic PVDF membrane can be used for lysate clarification, protein kinase assays, or bead- or resin-based separation assays.