Nunc® MicroWell™ 96-Well Tissue Culture Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
152039 152038
73521-012CS 330.86 USD
73521-012 73521-026
Nunc® MicroWell™ 96-Well Tissue Culture Plates, Thermo Scientific
Microplates Cell Culture Plates
These nonsterile polystyrene plates feature either Collagen I or Poly-D-Lysine coated surfaces for cell based assays
The coatings create a positive charge on the surface that ensures cell attachment, growth, and differentiation. Both plates have a flat bottom and feature a high-flange design. Plates are clear and have excellent optical clarity.
Ordering information: Both plates are supplied with lids. For replacement lids and sealing tapes see the 62409-118 series.