GNE-477 ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-(7-Methyl-6-((4-(methylsulfonyl)piperazin-1-yl)methyl)-4-morpholinothieno[3,2-d]pyrimidin-2-yl)pyrimidin-2-amine, 5-(7-Methyl-6-((4-(methylsulphonyl)piperazin-1-yl)methyl)-4-morpholinothieno[3,2-d]pyrimidin-2-yl)pyrimidin-2-amine

A975111-10MG A975111-1MG A975111-5MG
77618-682EA 70.04 USD
77618-682 77618-684 77618-686
GNE-477 ≥98%
GNE-477

Caution: for Research Use Only

Formula: C₂₁H₂₈N₈O₃S₂
MDL Number: MFCD20527726
CAS Number: 1032754-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR