(S)-1-(Boc-amino)-2-propanol ≥98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (S)-(2-hydroxypropyl)carbamate

A918594-1G A918594-5G
77880-745EA 17 USD
77880-745 77880-746
(S)-1-(Boc-amino)-2-propanol ≥98%
(S)-1-(Boc-amino)-2-propanol

Caution: for Research Use Only

Formula: C₈H₁₇NO₃
MW: 175.23 g/mol
MDL Number: MFCD04974340
CAS Number: 167938-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR