Dicarbonylcyclopentadienylcobalt 95% (by titrimetric analysis)
Supplier: Ambeed
|
Danger
|
Synonyms:
Cyclopentadienylcobalt dicarbonyl, Dicarbonylcyclopentadienylcobalt(I)
A915754-250MG
A915754-5G
77603-292EA
20.5
USD
77603-292
77627-752
Dicarbonylcyclopentadienylcobalt 95% (by titrimetric analysis)
Dicarbonylcyclopentadienylcobalt
Caution: for Research Use Only
Formula:
C₇H₅CoO₂ |
MDL Number:
MFCD00009589 CAS Number: 12078-25-0 UN: 1992 ADR: 3,6.1,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...