Sudan I ≥95% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: Scarlet B, 1-(Phenyldiazenyl)naphthalen-2-ol, 1-Phenylazo-2-naphthol, Oil Orange, Solvent Yellow 14, Sudan Yellow

A897815-100G A897815-25G
77614-610EA 16.48 USD
77614-610 77614-612
Sudan I ≥95% (by HPLC)
Sudan I

Caution: for Research Use Only

Formula: C₁₆H₁₂N₂O
MW: 248.28 g/mol
MDL Number: MFCD00003911
CAS Number: 842-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR