Adenosine 5'-monophosphate sodium salt hydrate ≥98%

Supplier: Ambeed
Warning

Synonyms: Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate hydrate(x:1:x), Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate hydrate, 5'-Adenylic acid,sodium salt, hydrate (9CI)

A853962-25G A853962-100G A853962-5G
77595-372EA 18.45 USD
77595-372 77603-160 77627-666
Adenosine 5'-monophosphate sodium salt hydrate ≥98%
Adenosine 5'-monophosphate sodium salt hydrate

Caution: for Research Use Only

Formula: C₁₀H₁₄N₅Na₂O₈P
MW: 409.200 g/mol
MDL Number: MFCD00065023
CAS Number: 149022-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR