Polyethylene glycol monostearate ≥98%
Supplier: Ambeed
|
Warning
|
A832813-1000G
A832813-100G
A832813-500G
77627-630EA
23.69
USD
77627-630
77627-632
77627-634
Polyethylene glycol monostearate ≥98%
Polyethylene glycol monostearate
Caution: for Research Use Only
Formula:
HO(CH₂CH₂O)nOCC₁₇H₃₅ |
MDL Number:
MFCD00148007 CAS Number: 9004-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...