Polypropylene glycol ~Mn ~ 400

Supplier: Ambeed
Warning

Synonyms: Poly(propylene oxide), PPG

A806185-100G A806185-25G
77670-146EA 8 USD
77670-146 77880-605
Polypropylene glycol ~Mn ~ 400
Polypropylene glycol

Caution: for Research Use Only

Formula: H(OCH₂CH₂CH₂)nOH
MW: 1025
MDL Number: MFCD00084446
CAS Number: 25322-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR