Coelenterazine h ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Dehydroxy)coelenterazine, CLZN-h, 2,8-dibenzyl-6-(4-hydroxyphenyl)imidazo[1,2-a]pyrazin-3(7H)-one, 2-Deoxycoelenterazine

A762072-10MG A762072-5MG
77627-516EA 166.86 USD
77627-516 77627-518
Coelenterazine h ≥98%
Coelenterazine h

Caution: for Research Use Only

Formula: C₂₆H₂₁N₃O₂
MW: 407.47 g/mol
MDL Number: MFCD00798394
CAS Number: 50909-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR