(4-(Triphenylsilyl)phenyl)boronic acid 95+%

Supplier: Ambeed
Warning

A754296-250MG A754296-5G
77886-708EA 43 USD
77886-708 77886-709
(4-(Triphenylsilyl)phenyl)boronic acid 95+%
Formula: C₂₄H₂₁BO₂Si
MDL Number: MFCD16619346
CAS Number: 852475-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR