Cellulose 97-102%, Technical Grade

Supplier: Ambeed
Warning

A751922-1000G A751922-100G
77602-924EA 69.7 USD
77602-924 77602-926
Cellulose 97-102%, Technical Grade
Cellulose

Caution: for Research Use Only

Formula: (C₁₂H₂₂O₁₁)n
MDL Number: MFCD00081512
CAS Number: 9004-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR