2-Fluoro-6-(trifluoromethyl)pyridine-3-boronic acid ≥95%
Supplier: Ambeed
|
Warning
|
A738458-100MG
A738458-1G
A738458-250MG
A738458-5G
77886-677EA
106
USD
77886-677
77886-678
77886-679
77886-680
2-Fluoro-6-(trifluoromethyl)pyridine-3-boronic acid ≥95%
2-Fluoro-6-(trifluoromethyl)pyridine-3-boronic acid
Caution: for Research Use Only
Formula:
C₆H₄BF₄NO₂ |
MDL Number:
MFCD12025974 CAS Number: 1150114-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...