Gramine ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(1H-Indol-3-yl)-N,N-dimethylmethanamine, 3-(Dimethylaminomethyl)-Indole

A719845-25G A719845-100G
77602-848EA 31.78 USD
77602-848 77673-972
Gramine ≥98%
Gramine

Caution: for Research Use Only

Formula: C₁₁H₁₄N₂
MW: 174.25 g/mol
MDL Number: MFCD00005629
CAS Number: 87-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR