Sakuranetin ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-5-Hydroxy-2-(4-hydroxyphenyl)-7-methoxychroman-4-one
A702347-10MG
A702347-1MG
A702347-5MG
77602-790EA
115.83
USD
77602-790
77602-792
77602-794
Sakuranetin ≥98%
Sakuranetin
Caution: for Research Use Only
Formula:
C₁₆H₁₄O₅ MW: 286.29 g/mol |
MDL Number:
MFCD00075651 CAS Number: 2957-21-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...