DDAO [7-hydroxy-9H-(1,3-dichloro-9,9-dimethylacridin-2-one)] ≥99%

Supplier: Ambeed
Warning

A698023-50MG A698023-100MG
77627-410EA 110.64 USD
77627-410 77880-447
DDAO [7-hydroxy-9H-(1,3-dichloro-9,9-dimethylacridin-2-one)] ≥99%
DDAO [7-hydroxy-9H-(1,3-dichloro-9,9-dimethylacridin-2-one)]

Caution: for Research Use Only

Formula: C₁₅H₁₁Cl₂NO₂
MDL Number: MFCD01311087
CAS Number: 118290-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR