(2-(Methoxycarbonyl)-4-methylphenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A660321-1G A660321-250MG A660321-5G
77886-561EA 218 USD
77886-561 77886-562 77886-563
(2-(Methoxycarbonyl)-4-methylphenyl)boronic acid ≥98%
(2-(Methoxycarbonyl)-4-methylphenyl)boronic acid

Caution: for Research Use Only

Formula: C₉H₁₁BO₄
MDL Number: MFCD08704671
CAS Number: 1256355-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR